Avantor® Hichrom HI-LAP, GC Columns
供应商: Avantor
证书
关于此项目
Application specifc column for lipids, sterols and triglycerides analysis.
Max temperature limits may change depending on the film thickness.
规格
- Catalog No:
- HIGCHI46-32-010-25
- HIGCHI46-25-010-25
- Phase name:
- HI-LAP
- HI-LAP
- Phase composition:
- Proprietary Phase developed for Lipid Analysis Phase, Saturated and Unsaturated Triglycerides, Sterols and Lipid analysis
- Proprietary Phase developed for Lipid Analysis Phase, Saturated and Unsaturated Triglycerides, Sterols and Lipid analysis
- Crossbond:
- Yes
- Yes
- 温度限值:
- 370 °C
- 370 °C