Order Entry
China
ContactUsLinkComponent
Bis(2-dimethylaminoethyl)disulphide dihydrochloride ≥98.0% (通过HPLC,滴定分析)
Irritant
Environment
Bis(2-dimethylaminoethyl)disulphide dihydrochloride ≥98.0% (通过HPLC,滴定分析)
Catalog # TCIAB2272-25G
Supplier:  TCI
CAS Number:  
Irritant
Environment
Bis(2-dimethylaminoethyl)disulphide dihydrochloride ≥98.0% (通过HPLC,滴定分析)
Catalog # TCIAB2272-25G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • 包装规格:
    25 g
  • 顺序:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :281.30 g/mol
EINECS:405-300-9
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:17339-60-5
Formula:C₈H₂₀N₂S₂
MDL Number:MFCD00190681