Specifications
- 浓度:10000 mg/L
- 基质:Nitric acid dil.
- 包装类型:Plastic bottle
- 包装规格:100 mL
- 元件:铊
- 符号:Tl
Specifications
About this item
Tl in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.