Iron standard solution, 1000 mg/L Fe in 0,5 M nitric acid (from Fe(NO₃)₃) Certipur® standard for AAS, Supelco®
Catalog # MERC1.19781.0100
Supplier: MERCK MILLIPORE
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- 浓度:1000 mg/L
- 基质:0,5 M HNO₃
- 包装类型:Plastic bottle
- 包装规格:100 mL
- M-Clarity quality level:MQ100
- 元件:铁
- 符号:Fe
Specifications
About this item
Certifications: Traceable to SRM from NIST.