Ground socket
NS 29/32
Catalog # BRND51467
Supplier: BRAND
Jump to:
Product Details & Documents
PTFE. With grip collar and sealing rings for excellent tightness.
- Greaseless seal will not contaminate sample
- Outstanding chemical resistance at operating temperatures from −200 to +260 °C
- Sturdy design for permanent use
Low leakage rate: Q < 1 · 10-4 mbar · l · s-1.
Specifications
- 用于:NS 29/32