MS calibration solution, API, sodium iodide and cesium iodide, Supelco®
Catalog # SUPL14379-30ML
Supplier: SUPELCO
CAS Number:
Specifications
- 包装规格:30 mL
- 说明:Sodium iodide (2 μg/μL) and cesium iodide (50 ng/μL) in 50/50 2-propanol/water
Specifications
About this item
Solution for LC-MS calibration.