LiChrospher® RP-18, HPLC Columns
: HICHROM
These LiChrospher® columns are silica available both endcapped and non-endcapped C18 phases. This feature enhances the chromatography selectivity options.
- Spherical porous silica
- High efficiency packed columns
- Catalog No:
- HICHLISPRP18-5250A
- HICHLISPRP185150AF
- HICHLISPRP18550AF
- HICHLISPRP18510C5S
- HICHLISPRP185300A
- HICHLISPRP18575AF
- HICHLISPRP185300AF
- HICHLISPRP18-510C5
- HICHLISPRP185250AM
- HICHLISPRP185250AF
- HICHLISPRP18-5125A
- HICHLISPRP185125AF
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- 碳负载:
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 21,0%
- 加端盖的:
- 否
- 否
- 否
- 否
- 否
- 否
- 否
- 否
- 否
- 否
- 否
- 否
- 表面积:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 环境认证:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)