ContactUsLinkComponent
pH buffer solutions, referenced at 20 °C, Fluka™
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # HONC33650-500ML
供应商:  HONEYWELL
CAS 编号:  
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # HONC33650-500ML
供应商:  HONEYWELL
CAS 编号:  
规格
  • 包装类型:
    Plastic bottle
  • pH缓冲类型:
    Borate
  • 包装规格:
    500 mL
  • 参考温度:
    20 °C
  • 顺序:
    62
  • pH精度:
    ±0.05
  • Pk Info:
    1×500 ml
  • pH 值:
    11,00