Specifications
- 范围:−50...+70 °C
- 分辨率:0,01 °C
- 说明:Dual-traceable thermometer with one probe
- 准确度:±0,3 °C
- 包装规格:1
Specifications
About this item
Traceable International Standards digital thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
- Stainless steel probe construction
- Monitor inlet and outlet simultaneously
- Visual and audible, high and low settable alarms
Absolute high-precision thermometer is ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater, and when using pH/conductivity standards.
Display shows minimum/maximum and current temperatures. MIN/MAX monitors readings overnight, on weekends, or for any time period. It displays the exact time and date when the MIN/MAX temperature occurred. Alarm provides two visual (LEDs) and two audio alerts when temperature rises above or falls below high and low set points. Unit displays the exact time and date when dual thermometer alarms are triggered. Alarms are programmable in 0.1° increments. Visual and audible alarms signal continuously (for days) even if temperature returns to non-alarm range. First thermometer ever designed to permit user one-button calibration.
Stainless-steel probe supplied with units shows the slightest temperature change. Probe is triple purpose for liquid, air/gas, and semisolids. Accurate readings with tip penetration of 8,46 mm. Probe has a diameter of 3,175 mm, stem length of 158,75 mm, overall length of 228,6 and a cable length of 1524 mm.