VWR®, Precision Weights, OIML, Class F1
Catalog # VWRU12766-374
Supplier: VWR BRAND PRIVATE LABEL USA
Specifications
- 重量:1 - 100 g
- 说明:Precision weight, with statement of accuracy
- 包装规格:1
- 权重数量:16
Specifications
About this item
These weights are perfectly flat on top so they can stack easily on center. The bottoms of the weights are slightly recessed to expose the smallest possible area to wear.
- Two-piece construction
- Cylindrical in shape with a tapered neck for easy gripping
They are manufactured from the finest stainless steel available including Alloy 8 stainless steel that has low magnetic permeability, uniform consistency and high resistance to corrosion.
Certifications: Conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111).