VWR® Performance, Refrigerators, for storage of flammable materials
证书
关于此项目
Designed for the refrigerated storage of flammable or volatile materials in general laboratory work areas.
- Data acquisition
- Data logging 24/7
- Digital temp chart (24-hour)
- Min/max temp history
- USB port for data transfer
No electrical components are located in the interior of the unit, and the electrical components of the compressor are sealed in a vapour-proof enclosure for additional safety. Units feature a digital microprocessor temperature controller that allows the precise temperature management necessary for critical samples and supplies.
Output: 4 to 20 mA
Certifications: Meet NFPA and OSHA guidelines 45 and 70. UL/C-UL listed.
规格
- Catalog No:
- VWRU10819-734
- VWRU10819-732
- 货号:
- VWRU10819-734
- VWRU10819-732
- 除霜:
- Cycle
- Cycle
- 货架/抽屉:
- 5 adjustable shelves
- 1 adjustable shelves, 3 fixed shelves
- 门型:
- 1 solid swing door
- 1 solid swing door
- 电气:
- 115 V, 60 Hz, 6 A
- 115 V, 60 Hz, 5 A
- 重量:
- 177,3 kg
- 147,8 kg