ContactUsLinkComponent
VWR®Traceable®, Digital timer/controller, three channels
VWR®Traceable®, Digital timer/controller, three channels
Catalog # VWRU23609-188
VWR®Traceable®, Digital timer/controller, three channels
Catalog # VWRU23609-188
Specifications
  • 说明:
    Lab control/timer, 3 channels
  • 宽度×深度×高度:
    120×165××89 mm
  • 准确度:
    0,01 %
  • 包装规格:
    1
Specifications
About this item
This Traceable® laboratory controller/timer combines all of the most frequently needed lab functions into a single, multi-purpose unit.
  • Calculator mode
  • Metric conversion
Unit tells time of day, has three separate alarm timing channels and converts lab measurements to metric units, and performs as a full-function calculator.
Program all three channels from 1 second to 100 hours. When time reaches zero, an audible electronic alarm and flashing signal serve as dual alerts for each channel. Calculator mode has floating decimal and six-digit LED display with 130 mm digits. The fourth channel is a clock that instantly displays time of day in hours, minutes, and seconds, with a.m./p.m. designation. Conversion factor provides instant and accurate answers on the most frequently used metric conversions, including weights, liquid measures, and temperatures.
Turns laboratory equipment on and off at any programmed time of day. Program it for up to 224 on/off switchings per day or 1568 per week. In countdown mode, it turns instruments on/off to the second. In repeat position, it cycles equipment on and off continuously at any selected interval.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery: Supplied with a one-year warranty and three-prong grounded plug.