VWR®Traceable®, Digital qc timer
Digital timer, 1 channels, 20 h, 0,01 %
目录编号 VWRU62344-902
Jump to:
Product Details & Documents
Simple three-key use and large digits for easy viewing.
- Count up/down
- Resolution is one minute
When countdown timing reaches zero, times up flashes on display, alarm sounds, and timer counts up showing time elapsed since alarm.
: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
: Supplied with battery, flip-open stand, clip, magnet, Traceable® certificate.
规格
- 说明:Digital timer, 1 channels, 20 h
- 准确度:0,01 %