容量瓶,透明玻璃,class A,带序列,带玻璃塞子
1000 ml, 0,3 ml, Volumetric flask
Catalog # VWRU10124-376
Supplier: VWR BRAND PRIVATE LABEL USA
Jump to:
Product Details & Documents
由3.3膨胀硼硅酸盐玻璃制成。
瓶身带序列,带Penny-head塞子。
Specifications
- 容量:1000 ml
- 说明:Volumetric flask
- 误差:0,3 ml