ContactUsLinkComponent
Glycan BEH amide columns, ACQUITY UPLC™
Glycan BEH amide columns, ACQUITY UPLC™
供应商:  WATERS
证书
Glycan BEH amide columns, ACQUITY UPLC™
供应商:  WATERS
证书
规格
  • Catalog No:
  • WATE186004741
  • WATE186004742
  • 碳负载:
  • 12%
  • 12%
  • pH 范围:
  • 2 - 11
  • 2 - 11
  • 表面积:
  • 185 m²/g
  • 185 m²/g
规格
相关信息