二乙二醇丁醚醋酸酯, J.T.Baker®
5162, Glass bottle, amber, narrow mouth, 4 L
Supplier: Avantor
Jump to:
Product Details & Documents
Synonyms: 2-(2-正丁氧基乙氧基)乙酸乙酯, 丁基二甘醇乙酸酯
- CAS Number:124-17-4
- Formula:C₁₀H₂₀O₄
- MDL Number:MFCD00009458
- Molecular Weight:204.27 g/mol
- Density:0,99 g/cm³
- Flash Pt:116,00 °C
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Identification (by IR):Passes Test
Specifications
- 包装类型:Glass bottle, amber, narrow mouth
- 包装规格:4 L
- 顺序:5162
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



