ContactUsLinkComponent
Suite of columns, Cogent TYPE-C™, method development kits
Suite of columns, Cogent TYPE-C™, method development kits
Catalog # MICS40100-10P-2
Supplier:  MICROSOLV
undefined
Suite of columns, Cogent TYPE-C™, method development kits
Catalog # MICS40100-10P-2
Supplier:  MICROSOLV
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • 列格式:
    Method development kit
  • 内径:
    2,10 mm
  • 颗粒大小:
    4 µm
  • 孔大小:
    100 Å
  • 长度:
    100 mm
  • 顺序:
    04
  • 包装规格:
    5
Specifications
About this item
The Cogent suite of columns is set of 5 columns of the same configuration consisting of one each of the Bidentate C18™, UDC-Cholesterol™, Bidentate C8™, Diamond Hydride™ and Silica C™.
  • Applicable in reverse phase, normal phase or aqueous normal phase
  • Kit provides an extremely wide spectrum of selectivies and possibilities
All of the columns in the suite can be used at low pH or high pH (upper limit is pH 8 for all except the C18 and the C8 columns which can be used up to pH 10,2).
All of the columns rapidly equilibrate between gradients or with initial use, making them very quick into service.