磷酸二氢钾, Millipore®
4024, Glass bottle for solids, 1 kg
Catalog # CALB529568-1
Supplier: CALBIOCHEM
Jump to:
Product Details & Documents
Synonyms: 磷酸一钾, 磷酸二氢钾, 无水磷酸二氢钾
- CAS Number:7778-77-0
- Formula:KH₂PO₄
- MDL Number:MFCD00011401
- Molecular Weight:136.09 g/mol
- Merck Index:13,07744
- Boiling Pt:∼100 °C (1013 hPa)
- Melting Pt:253 °C
- Density:2,3 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-913-4
- REACH:01-2119490224-41
Specifications
Specifications
- 包装类型:Glass bottle for solids
- 包装规格:1 kg
- 顺序:4024



