4-羟基苯甲酸甲酯
2500, Glass bottle for solids, 150 mg
供应商: EDQM
Jump to:
Product Details & Documents
同义词: 对羟基苯甲酸甲酯, 尼泊金甲酯
- CAS号:99-76-3
- 化学式:HOC₆H₄CO₂CH₃
- MDL 编号:MFCD00002352
- Molecular Weight:152.15 g/mol
- 默克索引:13,06129
- 沸点:270…280 °C (1013 hPa)
- 熔点:125…128 °C
- 密度:1,1208 g/cm³ (137 °C)
- 储存温度:Ambient
- EINECS:202-785-7
Documents
Certificates
按批号搜索规格
规格
- 包装类型:Glass bottle for solids
- 包装规格:150 mg
- 顺序:2500



