- 容量:1 ml
- 顺序:02
- 工作台重量:35 mg
- Phase:Diol (COHCOH)
- 包装规格:1
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
- Run times up to nine times faster than conventional SPE columns
- Unique laminar configuration
- Silica based
General applications: Non ionic, polar analytes.
These SPE columns can be used in normal phase mode.