ContactUsLinkComponent
J.T.Baker® BAKERBOND Speedisk® C18 Light Load, SPE Columns
J.T.Baker® BAKERBOND Speedisk® C18 Light Load, SPE Columns
目录编号 BAKR8151-00
供应商:  Avantor
J.T.Baker® BAKERBOND Speedisk® C18 Light Load, SPE Columns
目录编号 BAKR8151-00
供应商:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
规格
  • 容量:
    1 ml
  • 顺序:
    01
  • 工作台重量:
    10 mg
  • Phase:
    Octadecyl (C18) Light Load
  • 包装规格:
    1
规格
关于此项目
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Non ionic, non polar to polar analytes.
These SPE columns can be used in reversed phase mode.