DL-二硫苏糖醇
Marketsource
Supplier: BIO RAD
Jump to:
About this item
Synonyms: 1,4-二巯基苏糖醇, 克莱兰氏试剂, DTT
- CAS Number:3483-12-3
- Formula:C₄H₁₀O₂S₂
- MDL Number:MFCD00004877
- Molecular Weight:154.25 g/mol
- Boiling Pt:∼125 °C (1013 hPa)
- Melting Pt:42…44 °C
- Flash Pt:>110 °C
- Storage Temperature:Refrigerator
- EINECS:222-468-7
2 Options Available Below
Product Details & Documents
Synonyms: 1,4-二巯基苏糖醇, 克莱兰氏试剂, DTT
- CAS Number:3483-12-3
- Formula:C₄H₁₀O₂S₂
- MDL Number:MFCD00004877
- Molecular Weight:154.25 g/mol
- Boiling Pt:∼125 °C (1013 hPa)
- Melting Pt:42…44 °C
- Flash Pt:>110 °C
- Storage Temperature:Refrigerator
- EINECS:222-468-7
Product Family Options
Product Information
- 顺序包装类型包装规格AvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



