羟胺盐酸盐 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 盐酸羟胺
- CAS Number:5470-11-1
- Formula:NH₂OH·HCl
- MDL Number:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:13,04853
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:155…158 °C (dec.)
- Density:1,67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:226-798-2
- UN:2923
- ADR:8,III
3 Options Available Below
Product Details & Documents
Synonyms: 盐酸羟胺
- CAS Number:5470-11-1
- Formula:NH₂OH·HCl
- MDL Number:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:13,04853
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:155…158 °C (dec.)
- Density:1,67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:226-798-2
- UN:2923
- ADR:8,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Titration with NaOH:99.0 to 101.0 %
- Sulfate (SO4):≤0.005 %
- Iron (Fe):≤0.001 %
Product Family Options
Product Information
- 顺序包装类型包装规格AvailabilityPrice