Chromium standard solution, 1000 mg/L Cr in 0,5 M nitric acid (from Cr(NO₃)₃) Certipur® standard for AAS, Supelco®
目录编号 MERC1.19779.0100
供应商: MERCK MILLIPORE
CAS 编号:
规格
- 浓度:1000 mg/L
- 基质:0,5 M HNO₃
- 包装类型:Plastic bottle
- 包装规格:100 mL
- M-Clarity quality level:MQ100
- 元件:铬
- 符号:Cr
规格
关于此项目
Certifications: Traceable to SRM from NIST.