Vydac 214TP C4, HPLC columns
Supplier: HICHROM
Certificates
About this item
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins, haemoglobin variants, histones, insulin variants and membrane proteins.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Catalog No:
- HICH214GD52/N
- HICH214TP5215
- HICH214TP1010
- HICH214TP1022
- HICH214TP5115
- HICH214TP152022
- HICH214TP5210
- HICH214GK52/N
- HICH214TP10415
- HICH214TP510
- HICH214GK54/N
- HICH214GD104/N
- HICH214FSK1210
- HICH214TP104
- HICH214TP101015
- HICH214TP5410
- HICH214GD34/N
- HICH214GCC1210
- HICH214GD54/N
- HICH214GD51/N
- HICH214TP10152215
- HICH214TP5405
- HICH214GK104/N
- HICH214FSK1510
- HICH214TP53
- HICH214TP101510
- HICH214TP101522
- HICH214TP5415
- HICH214FSK510
- HICH214TP5315
- HICH214GCC1520/N
- HICH214GCC1010
- HICH214TP52
- HICH214TP5205
- HICH214GCC510
- HICH214TP51
- HICH214TP54
- HICH5138110
- HICH214GCC152010
- HICH214GK51/N
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- 碳负载:
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 加端盖的:
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 是
- 美国药典代码:
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- 环境认证:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)