Robotic tips for FLIPR Tetra, Axygen®
AXYGFT-384-C-R
: Corning
- 包装规格:50 racks, 384 tips
- 灭菌:-
- 容积:30 µl
- 顺序:02
- Environmentally Preferable:Low Manufacturing Impact
- 环境认证:I-RECs, ISO 14001 - Reynosa
- 尖端类型:FT-384-C-R
- 尖端颜色:透明
- 过滤器:-
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Designed for use in Molecular devices FLIPR tetra automation systems.
- Manufactured to stringent specifications under strict process controls
- Assembled via automation to ensure consistent performance and quality
- Certified RNase-/DNase-free and non pyrogenic
- Black and clear tips in 384-well format