Order Entry
China
ContactUsLinkComponent
Proparacaine Hydrochloride, Reference Standard
Proparacaine Hydrochloride, Reference Standard
目录编号 USPH1571001
供应商:  U.S. PHARMACOPEIA
CAS 编号:  
Proparacaine Hydrochloride, Reference Standard
目录编号 USPH1571001
供应商:  U.S. PHARMACOPEIA
CAS 编号:  

规格

  • 包装类型:
    Vial
  • 包装规格:
    200 mg
  • 标准:
    USP reference standard
  • CAS No.:
    5875-06-9
  • 说明:
    Proparacaine Hydrochloride
  • UN No.:
    2811
  • 公式:
    C₁₆H₂₇ClN₂O₃
  • 说明:
    Proparacaine Hydrochloride

规格

关于此项目

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard