dsDNA System, QuantiFluor®
02, QuantiFluor® dsDNA System, sufficient reagents for 2000 assays at a 200 µl scale, 1 mL
目录编号 PROME2670
供应商: PROMEGA
Jump to:
Product Details & Documents
The QuantiFluor® dsDNA System contains a fluorescent DNA-binding dye that enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA) in solution. The system is suitable for use in many molecular biology applications, including next-gen sequencing and other techniques where accurate DNA quantitation is important.
- Specific quantitation of dsDNA
- More sensitive than other methods for low-concentration samples
QuantiFluor® dsDNA dye shows minimal binding to single stranded DNA (ssDNA), RNA, protein and interfering compounds, allowing specific quantitation of dsDNA.
Delivery information: Kit includes QuantiFluor® dsDNA dye (1×1 ml), Lambda DNA standard (1×100 µg) and 20X TE buffer, pH 7,5 (1×25 ml).
规格
规格
- 包装规格:1 mL
- 说明:QuantiFluor® dsDNA System, sufficient reagents for 2000 assays at a 200 µl scale
- 顺序:02