ATP assay system, ENLITEN®
Catalog # PROMFF2000
Supplier: PROMEGA
CAS Number:
Specifications
- 包装规格:100 Assays
- 说明:ENLITEN® ATP Assay System
Specifications
About this item
The ENLITEN® ATP Assay System Bioluminescence Detection Kit is designed for the rapid and quantitative measurement of adenosine 5´-triphosphate (ATP) levels for the indirect detection of biocontamination.
- Fast and convenient assay method
- Easy to prepare reagents
- Less variation, stable light output
- Detects as little as 10⁻¹⁵ moles of ATP
The ENLITEN® ATP Assay System is applicable for the indirect measurement of bacteria, yeasts, fungi and other microorganisms in foodstuffs, beverages, water, woodpulp, cosmetics and other products. It can also be used for the assay of enzymes that produce or degrade ATP.
Delivery information: Kit contains rLuciferase/Luciferin reagent (1 vial), Reconstitution Buffer (12 ml), ATP-free water (25 ml) and ATP standard (1 vial).