dNTP mix
目录编号 PROMU1515
供应商: PROMEGA
CAS 编号:
规格
- 包装规格:1.000 µl
- 说明:dNTP mix, 10 mM
- 顺序:02
规格
关于此项目
dNTP Mix is a solution containing sodium salts of dATP, dCTP, dGTP and dTTP, each at 10 mM in water at pH 7,5. The total concentration of nucleotides is 40 mM. One microliter of the dNTP Mix in a 50 μl reaction will give a final dNTP concentration of 200 μM for each dNTP.
- Highly pure (>99% triphosphate)
- 10 mM each nucleotide in water
- Reduce pipetting steps and possibility of contamination by adding nucleotides as a mix