Vydac 218TP C18, HPLC columns
Supplier: HICHROM
Certificates
About this item
Vydac TP is the first range of wide pore media phases developed in the Vydac range and it is the industry's standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
- USP L1
First generation polymeric C18 phase with unique selectivity. Recommended for small polypeptides 4 - 5K MW, enzymatic digest fragments, natural and synthetic peptides, multi-ring compounds.
* Use with holder GCH10 and column coupler HI-081.
** Use with holder 80101/N and column coupler HI-081.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Catalog No:
- HICH218FSK510
- HICH218TP101522
- HICH218GCC510
- HICH218TP101510
- HICH218TP5305
- HICH218TP15202215
- HICH218TP5105
- HICH218GD34/N
- HICH218GD52/N
- HICH218GD54/N
- HICH218TP54
- HICH218GD51
- HICH218GK104/N
- HICH218TP53
- HICH218GCC1010
- HICH218GK32/N
- HICH218GK34/N
- HICH218TP152010
- HICH218TP152022
- HICH218TP510
- HICH218TP3215
- HICH218TP5115
- HICH218TP5410
- HICH218TP3410
- HICH218TP51010
- HICH218TP1022
- HICH218GCC1210
- HICH218TP3415
- HICH218TP5215
- HICH218TP104
- HICH218TP34
- HICH218TP1010
- HICH218TP3405
- HICH218GK52/N
- HICH218GK54/N
- HICH218TP5110
- HICH218TP5210
- HICH218TP5405
- HICH218GD104/N
- HICH218GD32/N
- HICH218TP5415
- HICH5119751
- HICH218TP52
- HICH218FSK1210
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- 美国药典代码:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- 环境认证:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)