Orders LinkContactUsLinkComponent
Buffer, Tris-EDTA solution
Tris-EDTA solution, clear, liquid, 1 L
目录编号 BDHB7732-1
供应商:   BDH US PRODUCED
Buffer, Tris-EDTA solution
目录编号 BDHB7732-1
供应商:  BDH US PRODUCED
CAS号:  
Buffer, Tris-EDTA solution
Tris-EDTA solution, clear, liquid, 1 L
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 1 升
Product Details & Documents
Tris-EDTA solution (TE solution) contains ultra pure deionised water, 0,5 M EDTA with pH 7,5 and 1 M tris with pH 8.
规格
规格
  • 包装规格:
    1 L
  • 说明:
    Tris-EDTA solution, clear, liquid
规格