Seal with Bore for Screw Caps, Silicone with PTFE Washer
LENZ01331406
- 孔径:6 mm
- 说明:Sealing ring, GL 14 thread, int. Ø 6 mm, ext. Ø 12 mm
- 材质:聚四氟乙烯/硅酮
- 顺序:01
- 包装规格:1
Silicone rubber sealing rings with PTFE washers.
- Temperature resistant from −200 to +250 °C
- Excellent chemical resistance
- Easy handling
For threaded connection caps with central bore and GL thread.