Support cushions for centrifuge bottles, Corning®
Supplier: CORNING SCIENCE PRODUCTS
Certificates
About this item
PP tubes, polyetherimide support cushions.
- Each case of 250 or 500 ml tubes contains a rack to facilitate handling
- Support cushions must be used unless the rotor has appropriately shaped V-bottom holders
- Tubes are sterile and certified non-pyrogenic
- Supplied with plug style caps
- Maximum RCF 6000×g
Ideal for applications requiring large-volume centrifugation.