VWR®, 试管口真空过滤系统
Supplier: VWR Collection
About this item
These units are ideal for minimising unnecessary transfers by filtering directly into the centrifuge tube.
- Sterilised by electron beam irradiation
- Non pyrogenic
PES膜:蛋白结合度低,低蛋白析出。非常适合于组织培养应用。
Delivery information: 每个PP离心管均配有一个单独包装的盖子用于储存。每个单独的组件都带有批号,以确保可追溯性。
Packaging: 包装在可重新密封的袋子中。离心管为单独包装。
Caution: 漏斗和试管的尺寸.
4 Options Available Below
Product Details & Documents
These units are ideal for minimising unnecessary transfers by filtering directly into the centrifuge tube.
- Sterilised by electron beam irradiation
- Non pyrogenic
PES膜:蛋白结合度低,低蛋白析出。非常适合于组织培养应用。
Delivery information: 每个PP离心管均配有一个单独包装的盖子用于储存。每个单独的组件都带有批号,以确保可追溯性。
Packaging: 包装在可重新密封的袋子中。离心管为单独包装。
Caution: 漏斗和试管的尺寸.
Specifications for Product Family
Specifications
- Catalog No:
- VWRI514-1071
- VWRI514-1070
- VWRI514-1069
- VWRI514-1068
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- 环境认证:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
完整的过滤单元
Product Information
- 顺序说明膜容量膜径孔大小AvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.VWRI514-1071Environmentally PreferableLow Manufacturing Impact环境认证ISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.VWRI514-1070Environmentally PreferableLow Manufacturing Impact环境认证ISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact
仅过滤杯
Product Information
- 顺序说明膜容量膜径孔大小AvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.VWRI514-1069Environmentally PreferableLow Manufacturing Impact环境认证ISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.VWRI514-1068Environmentally PreferableLow Manufacturing Impact环境认证ISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact
Centrifuge tube stands only
Product Information
- Catalog #说明颜色孔数量AvailabilityPrice
- For 1,5/2,0 ml microcentrifuge tubes and 15/50 ml centrifuge tubes绿色7Log in for availability1 * 5 件¥100.00