Handheld Digital Microscopes, Universal
Product Details & Documents
The Universal series provides a broad range of products with high image quality to satisfy the most demanding user. Magnification range: 10 to 220×.
- High speed USB 2.0 connection
- Metal or composite housing
- Edge series models with high quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
- Wireless streaming to a tablet, Smart phone, or computer is possible in combination with WF-10 unit (to be ordered separately)
- Integrated polariser greatly reduces glare on reflective surfaces (some models only)
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Models with Flexible LED Control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.
Delivery information: Supplied with protecting pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)) and calibration target.
Documents
规格
- 倍率:10 - 220×
- 型号:UNIVERSAL, AM7915MZT
- 分辨率:5,0 MP
- 说明:Handheld microscope with exchangeable caps + polariser, Edge
- 顺序:5
- 附加功能:AMR/EDOF/EDR/FLC
- 照明:8 LEDs