Measuring pipettes (Mohr), class A, reusable, PYREX®
Catalog # CORN7070-5
Supplier: CORNING SCIENCE PRODUCTS
Specifications
- 颜色代码:蓝色
- 容量:5,0 ml
- 长度:350 mm
- 顺序:03
- 分度:0,1 ml
- 包装规格:6
- 误差:±0,02 ml
Specifications
About this item
These reusable PYREX® glass pipettes are colour-coded by size for easy identification and sorting.
- Calibrated 'to deliver' (TD) their total capacity without blow-out
- Top end of 5 to 25 ml sizes is constricted (Mohr type)
- Coloured graduations are enameled onto the glass
- Colour-coded by size for easy identification
- Each pipette is individually serialised and certified
Certifications: Calibrated to Class A tolerances in accordance with ASTM E-542, ATSM E-694, and ASTM E-1293 specifications.
Delivery: Supplied with a certificate of identification and capacity, traceable to NIST standards.