WiFi temperature and humidity data loggers, new Saveris 2
Catalog # TEST0572.2034
Supplier: TESTO
Specifications
- 范围:T: −30…+50 °C
RH: 0 - 100 % - 分辨率:T: 0,1 °C
RH: 0,1 % - 数据存储器:2×10000
- 接口:WiFi
- 保护等级:IP 40
- 说明:Saveris 2-H1 with integrated temperature and humidity sensor
- 顺序:04
- 型号:testo Saveris 2 -H1
- 电池使用寿命:2 years
- 报警器:Time-controlled alarm, alarm delay
- 电池:4×AA batteries
- 测量时间:5 s - 24 hrs
- 记录间隔:1 min - 24 hrs
- 传感器:NTC int.
- 重量:240 g
- 宽度×深度×高度:95×75×30,5 mm
- 准确度:T: ±0,5 °C
RH: ±2 % - 包装规格:1
Specifications
About this item
The Saveris 2 WiFi data logger system is the modern solution for monitoring temperature and humidity value in storerooms and work rooms. Installing the system is child's play, and can be carried out either via a browser, or with the Saveris 2 app.
- Data transfer by wireless LAN
- All measurement data always available, anywhere, on any instrument
- Alarms when limit values are violated
- With Saveris 2 app for easier configuration, push alarms and WiFi range analysis
- Cost-free online data store (cloud)
- The long battery life additionally ensures that the Saveris 2 system needs to be serviced only rarely
The WiFi data loggers reliably record temperature, humidity at adjustable intervals and transmit the measurement values directly by wireless LAN to the cloud. The readings stored can be analyzed at any time, anywhere, using an internet-enabled smartphone, tablet or PC. Limit value violations are immediately reported by e-mail, optionally by SMS or via the Saveris 2 app as a push notification. This allows critical processes to be kept always under control, even if you are not on site.