J.T.Baker® BAKERBOND spe™ Phenyl, SPE Columns
Catalog # BAKR7095-03
Supplier: Avantor
Specifications
- 颗粒大小:40 µm
- 孔大小:60 Å
- 容量:3 ml
- 顺序:02
- 加端盖的:是
- 碳负载:10,6%
- 工作台重量:500 mg
- Phase:Phenyl
- 包装规格:50
Specifications
About this item
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
- Silica based
- Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
- Processing by vacuum or positive pressure
General applications: Polar from non polar/polar solvents using hydrogen bonding like mechanisms.
These SPE columns can be used in reversed phase mode.