SGE HT5 GC Capillary Columns
SGEA054668
- 列格式:Capillary
- 薄膜厚度:0,50 µm
- 内径:0,32 mm
- 长度:30 m
- 顺序:13
- Polarity:Low-polarity stationary phase
- 温度限值:400 °C
- Phase:HT5
The SGE HT5 GC capillary column provides a great low-polarity option for GC analyses. Recommended for: Simulated distillation, general hydrocarbon profiles, pesticides/herbicides, GCMS applications.
- Ultra high temperature columns
- Unique phase – no equivalent phases
- Ideal for simulated distillation applications (petroleum industry)
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.