SGE BPX-VOLATILES GC Capillary Columns
Catalog # SGEA054985
Supplier: Trajan Scientific and Medical
Specifications
- 列格式:Capillary
- 薄膜厚度:3 µm
- 内径:0,53 mm
- 长度:60 m
- 顺序:7
- Polarity:Mid-polarity stationary phase
- 温度限值:290 °C
- Phase:BPX-VOLATILES
Specifications
About this item
The SGE BPX-Volatiles GC Capillary Column provides a great polar option for GC analyses. Recommended for: Environmental analyses, volatile organics, alcohol analyses, USP G43, EPA 624, 502.2, SW-846 8240/8260.
- Polar phase
- EPA volatile organics analysis
- Able to be solvent rinsed
- Bonded and cross-linked
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.