Orders LinkContactUsLinkComponent
Vectashield® mounting medium
Vectashield® mounting medium
Vectashield® mounting medium, 10 mL
Catalog # VECTH-1000
Supplier:   VECTOR LABORATORIES
Vectashield® mounting medium
Catalog # VECTH-1000
CAS Number:  
Vectashield® mounting medium
Vectashield® mounting medium, 10 mL
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 毫升
Product Details & Documents
Vectashield® Mounting Medium is a unique, stable formula for preserving fluorescence. It is superior to other anti-fading reagents or mounting media for fluorescence microscopy. Some agents retard the rate of fading, but their initial application gives a severe reduction in emission. Others give a good initial reading but bleach upon illumination. Vectashield® Mounting Medium combines both advantages into one product. It provides strong initial fluorescence and retards photobleaching over time. Vectashield® Mounting Medium is supplied in a bottle fitted with a cone-seal cap. A cap fitted with a drop dispenser pipette is also included. The cone-seal cap can be replaced with the drop dispenser cap for routine use and during storage. Mounted slides should be stored at 4 °C. The fluorescence will remain for many weeks.
  • Prevents rapid loss of fluorescence during microscopic examination
  • Retains its anti-fading ability during long-term storage
  • Optically clear
  • Mounting medium to preserve fluorescence and counterstain DNA with DAPI (blue)
Documents
Specifications
Specifications
  • 包装规格:
    10 mL
  • 说明:
    Vectashield® mounting medium
Specifications