Lockout hasps
供应商: BRADY
Jump to:
关于此项目
These lockout hasps are ideal for use during group lockout situations.
- Hasps are constructed of vinyl-coated high tensile steel and are plated to resist rust
- Lock holes are 9,5 mm (³/₈") diameter to accommodate most standard lock shackles
2 Options Available Below
Product Details & Documents
These lockout hasps are ideal for use during group lockout situations.
- Hasps are constructed of vinyl-coated high tensile steel and are plated to resist rust
- Lock holes are 9,5 mm (³/₈") diameter to accommodate most standard lock shackles