Flasks, round bottom, glass, clear, standard ground joints, two necks
Catalog # CGLACG-1520-02
Supplier: CHEMGLASS
Specifications
- 容量:100 ml
- 顺序:01
- 包装规格:1
- 侧管颈:24/40 NS
- 中心颈:24/40 NS
Specifications
About this item
Borosilicate glass 3.3.
- Heavy wall
- Very good chemical resistance
- Very good temperature resistance
These flasks have standard taper outer joints on the both center neck and angled side neck.