Orders LinkContactUsLinkComponent
Rectangular semi-micro cuvettes
Rectangular semi-micro cuvettes
004, Semi-micro, 46 mm, 1,4 ml, 40 mm, Quartz (Suprasil®)
目录编号 HELL114F-10-40
供应商:   Hellma
Rectangular semi-micro cuvettes
目录编号 HELL114F-10-40
供应商:  Hellma
Rectangular semi-micro cuvettes
Rectangular semi-micro cuvettes
004, Semi-micro, 46 mm, 1,4 ml, 40 mm, Quartz (Suprasil®)
Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents
Rectangular semi-micro cuvettes with four polished windows and PTFE lids for fluorescence applications.
规格
  • 光学路径长度:
    40 mm
  • Type:
    Semi-micro
  • 容积:
    1,4 ml
  • 窗口材质:
    Quartz (Suprasil®)
  • 高度:
    46 mm
  • 顺序:
    004