ContactUsLinkComponent
Qualitative filter papers, wet strengthened grades, grade 1574 and 1574 ½, Whatman™
Qualitative filter papers, wet strengthened grades, grade 1574 and 1574 ½, Whatman™
Catalog # WHAT10314844
Supplier:  CYTIVA WHATMAN
undefined
Qualitative filter papers, wet strengthened grades, grade 1574 and 1574 ½, Whatman™
Catalog # WHAT10314844
Supplier:  CYTIVA WHATMAN
Specifications
  • 等级:
    1574 ½
  • 颗粒滞留:
    7 - 12 µm
  • 顺序:
    03
  • 尺寸:
    125 mm
  • 厚度:
    160 μm
  • 重量:
    90 g/m²
  • 过滤速度:
    85 s (Herzberg)
  • 包装规格:
    100
Specifications
About this item
Medium fast cellulose filter paper with high wet strength. This paper has the same chemical resistance characteristics as Grade 1573.
  • Folded filters for easy use in applications involving a conical filter funnel
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc.