Multicoated microplates
目录编号 CORN3829
Supplier: CORNING SCIENCE PRODUCTS
Specifications
- 建议工作容积:10 μl
- 灭菌:-
- 颜色:黑色/透明
- 说明:Flat bottom, low base, with lid
- 顺序:02
- 包装规格:10
- 孔体积:16,5 µl
Specifications
About this item
The Corning® multicoated microplate provides users access to six different surface treatments on a single plate.
This microplate is useful when determining the correct surface for assay requirements. Single surface microplates can then be used for the full screen or experiment. These multicoated plates are currently available in 96-well, 384-well, and 1536-well formats in black with clear bottom.