Mouse IgG2a Isotype Control [clone: eBM2a] (APC (Allophycocyanin))
Catalog # EBIO17-4724-42
Supplier: EBIOSCIENCE
Specifications
- 抗体类型:Control
- 抗原:Immunoglobulin G2 kappa
- 克隆形成能力:Monoclonal
- 克隆:eBM2a
- 结合:APC (Allophycocyanin)
- 流式细胞计:Yes
- 主机:Mouse
- 同位型:IgG2a kappa
- 形式:Liquid
- 存储缓冲:PBS, 0.2% BSA, 0.1% gelatin, 0.09% sodium azide, pH 7.2
- 存储温度:4° C, store in dark, DO NOT FREEZE!
- 浓度:5 µl/Test
- 净化:Affinity chromatography
- 抗体大小:100 Tests
- 包装规格:100 Tests
Specifications
About this item
Type: Control
Antigen: IgG2a κ
Clonality: Monoclonal
Clone: eBM2a
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: