Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- 抗体类型:Primary
- 抗原:Interferon gamma
- 克隆形成能力:Monoclonal
- 克隆:NIB42
- ELISA:Yes
- 主机:Mouse
- 同位型:IgG1 kappa
- 反应性:Human
- 中和:Yes
- 形式:Liquid
- 基因:3458
- 抗原同义词:Interferon gamma
- 存储缓冲:PBS, no preservative, pH 7.2
- 存储温度:4° C
- 浓度:1 mg/mL
- 净化:Affinity chromatography
- 抗体大小:50 µg
- 包装规格:50 µG
Type:
Antigen: IFNG
Clonality:
Clone: NIB42
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human