VWR® ARISTAR®, Boron Standard Solutions for ICP-MS
Catalog # VWRC85551.180
供应商: VWR Chemicals
CAS 编号:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
规格
- 浓度:10 ppm
- 基质:H₂O/tr.NH₄OH
- 包装类型:Plastic bottle
- 包装规格:100 mL
- 顺序:24
- 元件:硼
- 符号:B
规格
关于此项目
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0,5% (typical)
- Delivered with complete Certificate of Analysis
Certifications: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.